Assay Details
| Gene Symbol: Fas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007987.2 | NP_032013.2 | 4 - 5 | 484 | 57 | |
| GenBank mRNA | AK002590.1 | - | 4 - 5 | 435 | 57 | |
| BC061160.1 | - | 4 - 5 | 457 | 30302649 | 57 | |
| DQ846748.1 | - | 4 - 5 | 428 | 57 | ||
More Information
Set Membership: |
|

Fas
Genomic Map