Assay Details
| Gene Symbol: Lipa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111100.1 | NP_001104570.1 | 8 - 9 | 991 | 122 | |
| NM_021460.3 | NP_067435.3 | 8 - 9 | 995 | 122 | ||
| GenBank mRNA | AK088659.1 | - | 8 - 9 | 947 | 122 | |
| AK149791.1 | - | 8 - 9 | 955 | 122 | ||
| AK154804.1 | - | 8 - 9 | 937 | 120 | ||
| AK164007.1 | - | 8 - 9 | 991 | 122 | ||
| AK169567.1 | - | 8 - 9 | 961 | 122 | ||
| BC058564.1 | - | 8 - 9 | 943 | 6441278 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lipa
Genomic Map