| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900002917.mm.1 |
NM_026640.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002917.mm.1 |
NM_026640.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002917.mm.1 |
TC1900000247.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002918.mm.1 |
NM_026640.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002918.mm.1 |
NM_026640.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002918.mm.1 |
TC1900000247.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002916.mm.1 |
NM_026640.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002916.mm.1 |
NM_026640.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002916.mm.1 |
TC1900000247.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002919.mm.1 |
NM_026640.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002919.mm.1 |
NM_026640.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002919.mm.1 |
TC1900000247.mm.1 |
MTA TC |