Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900014334.mm.1 |
NM_001164363.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014334.mm.1 |
NM_001164363.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014334.mm.1 |
NM_001164365.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014334.mm.1 |
NM_001164365.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014334.mm.1 |
NM_029810.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014334.mm.1 |
NM_029810.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014334.mm.1 |
NR_028353.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014334.mm.1 |
NR_028353.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014334.mm.1 |
TC1900001617.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014336.mm.1 |
NM_001164363.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014336.mm.1 |
NM_001164363.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014336.mm.1 |
NM_001164365.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014336.mm.1 |
NM_001164365.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014336.mm.1 |
NM_029810.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014336.mm.1 |
NM_029810.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014336.mm.1 |
NR_028353.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014336.mm.1 |
NR_028353.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014336.mm.1 |
TC1900001617.mm.1 |
MTA TC |