Assay Details
| Gene Symbol: As3mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020577.2 | NP_065602.2 | 5 - 6 | 555 | 85 | |
| GenBank mRNA | AK002801.1 | - | 5 - 6 | 488 | 85 | |
| AK006571.1 | - | 5 - 6 | 540 | 85 | ||
| AK011320.1 | - | 5 - 6 | 507 | 85 | ||
| BC013468.1 | - | 5 - 6 | 480 | 85 | ||
| BC145663.1 | - | 5 - 6 | 497 | 40130966 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

As3mt
Genomic Map