Assay Details
Gene Symbol: Ddx51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027156.3 | NP_081432.2 | 13 - 14 | 1791 | 68 | |
GenBank mRNA | AK010015.1 | - | 4 - 5 | 395 | 68 | |
AK141361.1 | - | 13 - 14 | 1772 | 68 | ||
AK152599.1 | - | 13 - 14 | 1759 | 68 | ||
BC023700.1 | - | 10 - 11 | 1054 | 68 | ||
BC060646.1 | - | 13 - 14 | 1768 | 6853826 | 68 | |
BC069876.1 | - | 6 - 7 | 619 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
