Assay Details
Gene Symbol: Ddx51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027156.3 | NP_081432.2 | 7 - 8 | 1054 | 75 | |
GenBank mRNA | AK141361.1 | - | 7 - 8 | 1035 | 75 | |
AK152599.1 | - | 7 - 8 | 1022 | 75 | ||
BC023700.1 | - | 4 - 5 | 317 | 75 | ||
BC060646.1 | - | 7 - 8 | 1031 | 6853826 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
