Assay Details
Gene Symbol: Ddx51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027156.3 | NP_081432.2 | 9 - 10 | 1397 | 89 | |
GenBank mRNA | AK141361.1 | - | 9 - 10 | 1378 | 89 | |
AK152599.1 | - | 9 - 10 | 1365 | 89 | ||
BC023700.1 | - | 6 - 7 | 660 | 89 | ||
BC060646.1 | - | 9 - 10 | 1374 | 6853826 | 89 | |
BC069876.1 | - | 2 - 3 | 225 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
