Assay Details
| Gene Symbol: Noc4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153570.2 | NP_705798.2 | 7 - 8 | 783 | 63 | |
| GenBank mRNA | AK052601.1 | - | 7 - 8 | 783 | 63 | |
| AK088355.1 | - | 7 - 8 | 784 | 63 | ||
| AK146006.1 | - | 7 - 8 | 779 | 63 | ||
| AK172310.1 | - | 7 - 8 | 762 | 63 | ||
| BC024616.1 | - | 7 - 8 | 773 | 4218310 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Noc4l
Genomic Map