Assay Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009469.3 | NP_033495.2 | 9 - 10 | 1083 | 107 | |
| GenBank mRNA | AF053756.1 | - | 9 - 10 | 775 | 107 | |
| AF072370.1 | - | 9 - 10 | 766 | 107 | ||
| AK122357.1 | - | 8 - 9 | 585 | 107 | ||
| AK148317.1 | - | 9 - 10 | 1058 | 107 | ||
| AK154939.1 | - | 9 - 10 | 1100 | 107 | ||
| AK156534.1 | - | 9 - 10 | 1060 | 107 | ||
| BC053062.1 | - | 9 - 10 | 743 | 107 | ||
| BC057121.1 | - | 9 - 10 | 1083 | 6834534 | 107 | |
| BC059835.1 | - | 9 - 10 | 1034 | 6406755 | 107 | |
More Information
Set Membership: |
|

Ulk1
Genomic Map