Assay Details
Gene Symbol: Trim28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011588.3 | NP_035718.2 | 4 - 5 | 1291 | 66 | |
GenBank mRNA | AF230391.1 | - | 4 - 5 | 892 | 66 | |
AF230392.1 | - | 4 - 5 | 892 | 66 | ||
AK141357.1 | - | 4 - 5 | 1035 | 66 | ||
AK149995.1 | - | 1 - 2 | 116 | 66 | ||
BC020452.1 | - | 4 - 5 | 722 | 66 | ||
BC023335.1 | - | 4 - 5 | 1000 | 66 | ||
BC058391.1 | - | 4 - 5 | 737 | 6817165 | 66 | |
BC089337.1 | - | 4 - 5 | 1291 | 6811294 | 66 | |
U67303.1 | - | 4 - 5 | 1282 | 66 | ||
X99644.1 | - | 4 - 5 | 892 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
