Assay Details
| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027376.3 | NP_081652.2 | 10 - 11 | 1204 | 93 | |
| GenBank mRNA | AK033247.1 | - | 10 - 11 | 1191 | 93 | |
| AK134650.1 | - | 10 - 11 | 1204 | 93 | ||
| BC045601.1 | - | 7 - 8 | 2223 | 93 | ||
| CK624547.1 | - | 5 - 6 | 466 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
establishment of planar polarity
neural tube closure
hair follicle development
transport
multicellular organism development
negative regulation of cell proliferation
regulation of smoothened signaling pathway
embryonic body morphogenesis
protein transport
spinal cord dorsal/ventral patterning
cell projection organization
negative regulation of cell migration
nonmotile primary cilium assembly
cilium assembly
positive regulation of cilium assembly
embryonic skeletal system morphogenesis
negative regulation of canonical Wnt signaling pathway
negative regulation of neural crest formation
negative regulation of fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation
establishment of planar polarity
neural tube closure
hair follicle development
transport
multicellular organism development
negative regulation of cell proliferation
regulation of smoothened signaling pathway
embryonic body morphogenesis
protein transport
spinal cord dorsal/ventral patterning
cell projection organization
negative regulation of cell migration
nonmotile primary cilium assembly
cilium assembly
positive regulation of cilium assembly
embryonic skeletal system morphogenesis
negative regulation of canonical Wnt signaling pathway
negative regulation of neural crest formation
negative regulation of fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | BC045601.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | BC057127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | CO041869.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | AK134650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | AK033247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | BC045601.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | BC057127.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | CO041869.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | AK134650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P198019 | AK033247.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645673 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162351 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | BC045601.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | BC057127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | CO041869.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | AK134650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | AK033247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | BC045601.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | BC057127.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | CO041869.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | AK134650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P198019 | AK033247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P24206 | BC045601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P24206 | AK011195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P24206 | AK141792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | BC045601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | BC057127.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | CO041869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | AK134650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | AK033247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | BC045601.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | BC057127.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | CO041869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | AK134650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P198019 | AK033247.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06307711 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06307696 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06657221 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06307692 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06307709 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06307707 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06362611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25089991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06307711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06216523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06362611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06307701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06657220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25089989 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552786 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552795 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552782 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552784 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562964 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552781 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552790 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552785 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552789 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552788 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552783 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552780 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552787 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438749_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431025_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552779 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562963 | - | - |
| Mouse Genome 430 2.0 Array | 1431025_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431025_at | - | - |
| Mouse Genome 430 2.0 Array | 1438749_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438749_at | - | - |
| Mouse Genome 430 2.0 Array | 1447494_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447494_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70300_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70300_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700006949.mm.1 | TC0700000821.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006949.mm.1 | NM_027376.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006949.mm.1 | NM_027376.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006950.mm.1 | TC0700000821.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006950.mm.1 | NM_027376.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006950.mm.1 | NM_027376.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006954.mm.1 | TC0700000821.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006954.mm.1 | NM_027376.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006954.mm.1 | NM_027376.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006953.mm.1 | TC0700000821.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700006953.mm.1 | NM_027376.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700006953.mm.1 | NM_027376.3 | Refseq |
| Mouse Expression Set 430 | 1438749_PM_at | - | - |
| Mouse Expression Set 430 | 1431025_PM_at | - | - |
| Mouse Expression Set 430 | 1438749_at | - | - |
| Mouse Expression Set 430 | 1431025_at | - | - |
| Murine Genome U74v2 Set | 140894_PM_at | - | - |
| Murine Genome U74v2 Set | 137588_r_at | - | - |
| Murine Genome U74v2 Set | 140894_at | - | - |
| Murine Genome U74v2 Set | 137588_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431025_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438749_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438749_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431025_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 491436 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 106550070 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239948 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239948 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236746 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106550070 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106550070 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236746 | - | - |

Fuz
Genomic Map