Assay Details
| Gene Symbol: Fuz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027376.3 | NP_081652.2 | 7 - 8 | 959 | 90 | |
| GenBank mRNA | AK033247.1 | - | 7 - 8 | 946 | 90 | |
| AK035030.1 | - | 692 | 90 | |||
| AK045745.1 | - | 954 | 90 | |||
| AK134650.1 | - | 7 - 8 | 959 | 90 | ||
| AK168939.1 | - | 7 - 8 | 941 | 90 | ||
| BC045601.1 | - | 6 - 7 | 1823 | 90 | ||
| BC057127.1 | - | 1 - 2 | 605 | 90 | ||
| CK624547.1 | - | 2 - 3 | 221 | 90 | ||
| CO427180.1 | - | 5 - 6 | 532 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fuz
Genomic Map