Assay Details
| Gene Symbol: Fcgrt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010189.3 | NP_034319.2 | 4 - 5 | 994 | 53 | |
| GenBank mRNA | AK152879.1 | - | 4 - 5 | 930 | 53 | |
| BC003786.1 | - | 4 - 5 | 929 | 3485649 | 53 | |
| D37874.1 | - | 4 - 5 | 927 | 53 | ||
| L17022.1 | - | 3 - 4 | 633 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcgrt
Genomic Map