Assay Details
| Gene Symbol: Prrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022999.1 | NP_075375.1 | 2 - 3 | 218 | 81 | |
| GenBank mRNA | AK005453.1 | - | 3 - 4 | 262 | 81 | |
| BC025098.1 | - | 2 - 3 | 218 | 4159863 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prrg2
Genomic Map