Assay Details
| Gene Symbol: Prrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022999.1 | NP_075375.1 | 4 - 5 | 441 | 72 | |
| GenBank mRNA | AK005453.1 | - | 5 - 6 | 484 | 72 | |
| BC025098.1 | - | 4 - 5 | 441 | 4159863 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prrg2
Genomic Map