Assay Details
| Gene Symbol: Ap2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077264.1 | NP_001070732.1 | 16 - 17 | 2321 | 69 | |
| NM_007458.2 | NP_031484.1 | 17 - 18 | 2387 | 69 | ||
| GenBank mRNA | AK135424.1 | - | 16 - 17 | 2250 | 69 | |
| AK155515.1 | - | 16 - 17 | 2258 | 69 | ||
| BC031433.1 | - | 17 - 18 | 2337 | 4481643 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap2a1
Genomic Map