Assay Details
| Gene Symbol: Snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005045.2 | - | 1 - 2 | 187 | 72 | |
| AK011564.1 | - | 1 - 2 | 143 | 72 | ||
| AK077425.1 | - | 1 - 2 | 189 | 72 | ||
| AK133115.1 | - | 1 - 2 | 188 | 72 | ||
| AK146729.1 | - | 1 - 2 | 188 | 72 | ||
| AK148013.1 | - | 1 - 2 | 214 | 72 | ||
| BC049128.1 | - | 1 - 2 | 153 | 72 | ||
| BC131994.1 | - | 1 - 2 | 80 | 40130271 | 72 | |
| BC132020.1 | - | 1 - 2 | 80 | 40130297 | 72 | |
| BY297843.1 | - | 1 - 2 | 237 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011564.1, AK077425.1, AK133115.1, AK146729.1, BC049128.1, BC131994.1, BC132020.1 transcripts.
Set Membership: |
|

Snrnp70
Genomic Map