Assay Details
| Gene Symbol: Snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011564.1 | - | 7 - 8 | 623 | 85 | |
| AK014311.2 | - | 7 - 8 | 664 | 85 | ||
| AK133115.1 | - | 7 - 8 | 668 | 85 | ||
| AK146729.1 | - | 7 - 8 | 668 | 85 | ||
| AK148013.1 | - | 7 - 8 | 694 | 85 | ||
| BC131994.1 | - | 7 - 8 | 560 | 40130271 | 85 | |
| BC132020.1 | - | 7 - 8 | 560 | 40130297 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snrnp70
Genomic Map