Assay Details
| Gene Symbol: Rac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009007.2 | NP_033033.1 | 3 - 4 | 420 | 66 | |
| GenBank mRNA | AK008435.1 | - | 3 - 4 | 421 | 66 | |
| AK009017.1 | - | 3 - 4 | 421 | 66 | ||
| AK011072.2 | - | 3 - 4 | 421 | 66 | ||
| AK034601.1 | - | 3 - 4 | 421 | 66 | ||
| AK047969.1 | - | 3 - 4 | 423 | 66 | ||
| AK088825.1 | - | 3 - 4 | 432 | 66 | ||
| BC003828.1 | - | 3 - 4 | 421 | 3593957 | 66 | |
| BC051053.1 | - | 3 - 4 | 420 | 5038182 | 66 | |
| CX242948.1 | - | 3 - 4 | 457 | 66 | ||
| JN971018.1 | - | 3 - 4 | 224 | 66 | ||
| X57277.1 | - | 3 - 4 | 364 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rac1
Genomic Map