Assay Details
| Gene Symbol: Actb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK075973.1 | - | 2 - 3 | 200 | 72 | |
| AK078935.1 | - | 2 - 3 | 203 | 72 | ||
| AK088691.1 | - | 2 - 3 | 200 | 72 | ||
| AK145191.1 | - | 2 - 3 | 203 | 72 | ||
| AK145196.1 | - | 2 - 3 | 203 | 72 | ||
| AK145308.1 | - | 2 - 3 | 203 | 72 | ||
| AK147787.1 | - | 2 - 3 | 200 | 72 | ||
| AK149920.1 | - | 2 - 3 | 203 | 72 | ||
| AK150545.1 | - | 2 - 3 | 204 | 72 | ||
| AK150553.1 | - | 2 - 3 | 204 | 72 | ||
| AK150558.1 | - | 2 - 3 | 204 | 72 | ||
| AK150570.1 | - | 2 - 3 | 204 | 72 | ||
| AK150662.1 | - | 2 - 3 | 201 | 72 | ||
| AK150711.1 | - | 2 - 3 | 203 | 72 | ||
| AK150854.1 | - | 2 - 3 | 204 | 72 | ||
| AK150866.1 | - | 2 - 3 | 203 | 72 | ||
| AK150879.1 | - | 2 - 3 | 203 | 72 | ||
| AK151010.1 | - | 2 - 3 | 200 | 72 | ||
| AK151136.1 | - | 2 - 3 | 203 | 72 | ||
| AK151145.1 | - | 2 - 3 | 200 | 72 | ||
| AK151159.1 | - | 2 - 3 | 203 | 72 | ||
| AK151166.1 | - | 2 - 3 | 200 | 72 | ||
| AK151190.1 | - | 2 - 3 | 203 | 72 | ||
| AK151202.1 | - | 2 - 3 | 203 | 72 | ||
| AK151226.1 | - | 2 - 3 | 200 | 72 | ||
| AK151277.1 | - | 2 - 3 | 200 | 72 | ||
| AK151297.1 | - | 2 - 3 | 203 | 72 | ||
| AK151350.1 | - | 2 - 3 | 200 | 72 | ||
| AK151385.1 | - | 2 - 3 | 203 | 72 | ||
| AK151386.1 | - | 2 - 3 | 200 | 72 | ||
| AK151398.1 | - | 2 - 3 | 203 | 72 | ||
| AK151444.1 | - | 2 - 3 | 203 | 72 | ||
| AK151468.1 | - | 2 - 3 | 200 | 72 | ||
| AK151663.1 | - | 2 - 3 | 203 | 72 | ||
| AK151712.1 | - | 2 - 3 | 204 | 72 | ||
| AK151898.1 | - | 2 - 3 | 201 | 72 | ||
| AK151956.1 | - | 2 - 3 | 200 | 72 | ||
| AK151995.1 | - | 2 - 3 | 203 | 72 | ||
| AK151999.1 | - | 2 - 3 | 203 | 72 | ||
| AK152043.1 | - | 2 - 3 | 204 | 72 | ||
| AK152052.1 | - | 2 - 3 | 201 | 72 | ||
| AK152083.1 | - | 2 - 3 | 204 | 72 | ||
| AK152218.1 | - | 2 - 3 | 204 | 72 | ||
| AK152433.1 | - | 2 - 3 | 201 | 72 | ||
| AK152470.1 | - | 2 - 3 | 201 | 72 | ||
| AK152591.1 | - | 2 - 3 | 204 | 72 | ||
| AK152615.1 | - | 2 - 3 | 203 | 72 | ||
| AK152651.1 | - | 2 - 3 | 203 | 72 | ||
| AK152671.1 | - | 2 - 3 | 203 | 72 | ||
| AK152813.1 | - | 2 - 3 | 204 | 72 | ||
| AK152841.1 | - | 2 - 3 | 203 | 72 | ||
| AK152844.1 | - | 2 - 3 | 200 | 72 | ||
| AK153461.1 | - | 2 - 3 | 200 | 72 | ||
| AK159759.1 | - | 2 - 3 | 203 | 72 | ||
| AK159834.1 | - | 2 - 3 | 200 | 72 | ||
| AK160029.1 | - | 2 - 3 | 200 | 72 | ||
| AK166349.1 | - | 2 - 3 | 203 | 72 | ||
| AK167117.1 | - | 2 - 3 | 203 | 72 | ||
| AK167825.1 | - | 2 - 3 | 203 | 72 | ||
| BC138611.1 | - | 2 - 3 | 168 | 8861631 | 72 | |
| BC138614.1 | - | 2 - 3 | 168 | 8861634 | 72 | |
| BY783897.1 | - | 2 - 3 | 231 | 72 | ||
| EF095208.1 | - | 1 - 2 | 92 | 72 | ||
| J04181.1 | - | 2 - 3 | 184 | 72 | ||
| X03672.1 | - | 2 - 3 | 202 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Actb
Genomic Map