Assay Details
| Gene Symbol: Fbxl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033312.3 | NP_001028484.2 | 1 - 2 | 133 | 82 | |
| GenBank mRNA | AK049302.1 | - | 1 - 2 | 86 | 82 | |
| AK144757.1 | - | 1 - 2 | 134 | 82 | ||
| AK147353.1 | - | 1 - 2 | 317 | 82 | ||
| AK154354.1 | - | 1 - 2 | 113 | 82 | ||
| BC115599.2 | - | 1 - 2 | 105 | 82 | ||
| BC139739.1 | - | 1 - 2 | 105 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154354.1, BC115599.2, BC139739.1, NM_001033312.3 transcripts.
Set Membership: |
|

Fbxl18
Genomic Map