Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500004373.mm.1 |
NM_033371.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004373.mm.1 |
NM_033371.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004373.mm.1 |
TC1500000604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004370.mm.1 |
NM_033371.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004370.mm.1 |
NM_033371.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004370.mm.1 |
TC1500000604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004366.mm.1 |
NM_033371.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004366.mm.1 |
NM_033371.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004366.mm.1 |
TC1500000604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004369.mm.1 |
NM_033371.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004369.mm.1 |
NM_033371.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004369.mm.1 |
TC1500000604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004371.mm.1 |
NM_033371.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004371.mm.1 |
NM_033371.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004371.mm.1 |
TC1500000604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004372.mm.1 |
NM_033371.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004372.mm.1 |
NM_033371.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004372.mm.1 |
TC1500000604.mm.1 |
MTA TC |