Assay Details
Gene Symbol: Ppp1r16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033371.2 | NP_203535.1 | 2 - 3 | 1066 | 54 | |
GenBank mRNA | AK154865.1 | - | 2 - 3 | 1051 | 54 | |
AK171452.1 | - | 2 - 3 | 1051 | 54 | ||
AK220248.1 | - | 2 - 3 | 1066 | 54 | ||
AY010723.1 | - | 1 - 2 | 261 | 54 | ||
BC033318.1 | - | 1 - 2 | 626 | 54 | ||
BC044775.1 | - | 1 - 2 | 748 | 54 | ||
BC049958.1 | - | 1 - 2 | 748 | 54 | ||
BC057450.1 | - | 1 - 2 | 683 | 3966334 | 54 | |
BC096620.1 | - | 2 - 3 | 1066 | 6390076 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
