Assay Details
Gene Symbol: Ppp1r16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033371.2 | NP_203535.1 | 7 - 8 | 1638 | 58 | |
GenBank mRNA | AK150235.1 | - | 4 - 5 | 433 | 58 | |
AK154865.1 | - | 7 - 8 | 1623 | 58 | ||
AK171452.1 | - | 7 - 8 | 1623 | 58 | ||
AY010723.1 | - | 6 - 7 | 833 | 58 | ||
BC044775.1 | - | 5 - 6 | 1393 | 58 | ||
BC049958.1 | - | 5 - 6 | 1393 | 58 | ||
BC057450.1 | - | 6 - 7 | 1255 | 3966334 | 58 | |
BC096620.1 | - | 7 - 8 | 1638 | 6390076 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
