Assay Details
| Gene Symbol: Ppp1r16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033371.2 | NP_203535.1 | 8 - 9 | 1715 | 74 | |
| GenBank mRNA | AK150235.1 | - | 5 - 6 | 510 | 74 | |
| AK154865.1 | - | 8 - 9 | 1700 | 74 | ||
| AK171452.1 | - | 8 - 9 | 1700 | 74 | ||
| AK181600.1 | - | 2 - 3 | 133 | 74 | ||
| AY010723.1 | - | 7 - 8 | 910 | 74 | ||
| BC044775.1 | - | 6 - 7 | 1470 | 74 | ||
| BC049958.1 | - | 6 - 7 | 1470 | 74 | ||
| BC057450.1 | - | 7 - 8 | 1332 | 3966334 | 74 | |
| BC096620.1 | - | 8 - 9 | 1715 | 6390076 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r16a
Genomic Map