Assay Details
| Gene Symbol: Adap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172723.4 | NP_766311.2 | 2 - 3 | 315 | 61 | |
| GenBank mRNA | BC056934.1 | - | 1 - 2 | 41 | 61 | |
| BC079634.1 | - | 1 - 2 | 109 | 61 | ||
| CX242691.1 | - | 2 - 3 | 315 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adap1
Genomic Map