Assay Details
| Gene Symbol: Cox19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197980.1 | NP_932097.1 | 2 - 3 | 208 | 135 | |
| GenBank mRNA | AK002486.1 | - | 2 - 3 | 224 | 135 | |
| AK003697.1 | - | 2 - 3 | 252 | 135 | ||
| AK013260.1 | - | 2 - 3 | 224 | 135 | ||
| AK013289.1 | - | 2 - 3 | 252 | 135 | ||
| AK148526.1 | - | 2 - 3 | 181 | 135 | ||
| BC031792.1 | - | 2 - 3 | 208 | 4193998 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135005 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24134880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135004 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | AK148526.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | BC031792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | AK013289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | NM_197980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | AK002486.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | AK148526.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | BC031792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | AK013289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | NM_197980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177341 | AK002486.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182890 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399545 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898874 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493575 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23429 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122042 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | AK148526.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | BC031792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | AK013289.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | NM_197980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | AK002486.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | AK148526.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | BC031792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | AK013289.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | NM_197980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177341 | AK002486.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24134996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24134997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24134994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24134990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24134995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24134992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24134993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24135026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24134999 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05765045 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122042 | - | - |
| Whole Mouse Genome G4122A | A_52_P177341 | AK148526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177341 | BC031792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177341 | AK013289.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177341 | NM_197980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177341 | AK002486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177341 | AK148526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177341 | BC031792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177341 | AK013289.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177341 | NM_197980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177341 | AK002486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413687 | AK148526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413687 | AK013289.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109360 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24134880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135005 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24135004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24135019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24134984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04566235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04464469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04210195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04421469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04566236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04977467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04722937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04896965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04748464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04458382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04888424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04718258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04464482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04442083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24135039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04442081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04207454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04805653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24134985 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535083 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434923_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535080 | - | - |
| Mouse Genome 430 2.0 Array | 1434923_at | - | - |
| Mouse Genome 430 2.0 Array | 1434923_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68033_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68033_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500031930.mm.1 | TC0500003489.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031930.mm.1 | NM_197980.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031930.mm.1 | NM_197980.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031931.mm.1 | TC0500003489.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031931.mm.1 | NM_197980.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031931.mm.1 | NM_197980.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031929.mm.1 | TC0500003489.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031929.mm.1 | NM_197980.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031929.mm.1 | NM_197980.1 | Refseq |
| Mouse Expression Set 430 | 1434923_PM_at | - | - |
| Mouse Expression Set 430 | 1434923_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434923_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434923_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434923_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434923_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 784384 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4280575 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280575 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221936 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221936 | - | - |
| mouse-6 v1.1 (Illumina) | scl0068033.1_52-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280575 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0068033.1_52-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280575 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221936 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221936 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0068033.1_52-S | - | - |

Cox19
Genomic Map