Assay Details
| Gene Symbol: Prkar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JF720024.1 | - | 1 - 2 | 196 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the JF720024.1 transcripts.
Set Membership: |
|

Prkar1b
Genomic Map