Assay Details
| Gene Symbol: Ankle2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253814.1 | NP_001240743.1 | 6 - 7 | 1605 | 99 | |
| NM_027922.2 | NP_082198.1 | 6 - 7 | 1602 | 99 | ||
| GenBank mRNA | AK003234.1 | - | 3 - 4 | 404 | 99 | |
| AK129192.1 | - | 5 - 6 | 931 | 99 | ||
| AK132187.1 | - | 6 - 7 | 1607 | 99 | ||
| AK142497.1 | - | 4 - 5 | 654 | 99 | ||
| BC065071.1 | - | 6 - 7 | 1565 | 6410033 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankle2
Genomic Map