Assay Details
| Gene Symbol: Smpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195537.1 | NP_001182466.1 | 5 - 6 | 1228 | 52 | |
| GenBank mRNA | AK005822.1 | - | 4 - 5 | 510 | 52 | |
| BY705974.1 | - | 4 - 5 | 507 | 52 | ||
| CF197814.1 | - | 3 - 4 | 304 | 52 | ||
| GU144514.1 | - | 5 - 6 | 1228 | 52 | ||
More Information
Set Membership: |
|

Smpd5
Genomic Map