Assay Details
| Gene Symbol: Gpihbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026730.1 | NP_081006.1 | 3 - 4 | 314 | 75 | |
| GenBank mRNA | AB095543.1 | - | 3 - 4 | 321 | 75 | |
| AK003305.1 | - | 3 - 4 | 330 | 75 | ||
| BC061225.1 | - | 3 - 4 | 314 | 30298145 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transport
lipid transport
intracellular protein transport
protein import
cholesterol transport
protein localization to cell surface
cholesterol homeostasis
transcytosis
protein stabilization
positive regulation of lipoprotein lipase activity
triglyceride homeostasis
protein transmembrane transport
positive regulation of chylomicron remodeling
positive regulation of chylomicron remnant clearance
high-density lipoprotein particle binding
lipid binding
protein transmembrane transporter activity
lipase binding
chylomicron binding
lipoprotein particle binding
transport
lipid transport
intracellular protein transport
protein import
cholesterol transport
protein localization to cell surface
cholesterol homeostasis
transcytosis
protein stabilization
positive regulation of lipoprotein lipase activity
triglyceride homeostasis
protein transmembrane transport
positive regulation of chylomicron remodeling
positive regulation of chylomicron remnant clearance
high-density lipoprotein particle binding
lipid binding
protein transmembrane transporter activity
lipase binding
chylomicron binding
lipoprotein particle binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331328 | BC061225.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331328 | NM_026730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331328 | AB095543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331328 | AK003305.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331328 | BC061225.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331328 | NM_026730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331328 | AB095543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P331328 | AK003305.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342725 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148572 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331328 | BC061225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331328 | NM_026730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331328 | AB095543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331328 | AK003305.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331328 | BC061225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331328 | NM_026730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331328 | AB095543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P331328 | AK003305.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P331328 | BC061225.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P331328 | NM_026730.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P331328 | AB095543.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P331328 | AK003305.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P331328 | BC061225.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P331328 | NM_026730.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P331328 | AB095543.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P331328 | AK003305.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342837 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148572 | - | - |
| Whole Mouse Genome G4122A | A_51_P331328 | BC061225.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331328 | NM_026730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331328 | AB095543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331328 | AK003305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331328 | BC061225.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331328 | NM_026730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331328 | AB095543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P331328 | AK003305.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148572 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12280335 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12280318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12280299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12280302 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12280329 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12280317 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12280316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11909196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12280304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12213557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11909200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12209642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11909199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12280335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12263584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12213538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12280311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12280329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11896992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12209641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12342518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12263583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12294575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12213566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342825 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10424696 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424698 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424699 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424697 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453022_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10424695 | - | - |
| Mouse Genome 430 2.0 Array | 1453022_at | - | - |
| Mouse Genome 430 2.0 Array | 1453022_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68453_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68453_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500003804.mm.1 | NM_026730.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003804.mm.1 | NM_026730.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500003804.mm.1 | TC1500000557.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453022_at | - | - |
| Mouse Expression Set 430 | 1453022_PM_at | - | - |
| Murine Genome U74v2 Set | 165950_PM_r_at | - | - |
| Murine Genome U74v2 Set | 165950_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453022_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453022_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3060739 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2614842 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217969 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2614842 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217969 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1217969 | - | - |

Gpihbp1
Genomic Map