Assay Details
| Gene Symbol: Top1mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028404.2 | NP_082680.1 | 12 - 13 | 1571 | 94 | |
| GenBank mRNA | AF362952.1 | - | 12 - 13 | 1572 | 94 | |
| AK013665.1 | - | 6 - 7 | 706 | 94 | ||
| AK147119.1 | - | 12 - 13 | 1574 | 94 | ||
| BC138582.1 | - | 12 - 13 | 1563 | 8861602 | 94 | |
| BC138583.1 | - | 12 - 13 | 1563 | 8861603 | 94 | |
| BC145336.1 | - | 12 - 13 | 1560 | 9053892 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top1mt
Genomic Map