Assay Details
| Gene Symbol: Top1mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028404.2 | NP_082680.1 | 3 - 4 | 378 | 88 | |
| GenBank mRNA | AF362952.1 | - | 3 - 4 | 379 | 88 | |
| AK147119.1 | - | 3 - 4 | 381 | 88 | ||
| BC138582.1 | - | 3 - 4 | 370 | 8861602 | 88 | |
| BC138583.1 | - | 3 - 4 | 370 | 8861603 | 88 | |
| BC145336.1 | - | 3 - 4 | 370 | 9053892 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top1mt
Genomic Map