Assay Details
| Gene Symbol: Top1mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028404.2 | NP_082680.1 | 4 - 5 | 496 | 98 | |
| GenBank mRNA | AF362952.1 | - | 4 - 5 | 497 | 98 | |
| AK147119.1 | - | 4 - 5 | 499 | 98 | ||
| BC138582.1 | - | 4 - 5 | 488 | 8861602 | 98 | |
| BC138583.1 | - | 4 - 5 | 488 | 8861603 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top1mt
Genomic Map