Assay Details
| Gene Symbol: 9030619P08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108041.1 | 2 - 3 | 209 | 71 | ||
| GenBank mRNA | AK136484.1 | - | 2 - 3 | 120 | 71 | |
| BC087965.1 | - | 2 - 3 | 209 | 71 | ||
| BC147461.1 | - | 2 - 3 | 111 | 9056103 | 71 | |
| BC147464.1 | - | 2 - 3 | 111 | 9056106 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122086 | AK136484.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122086 | NM_001039720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122086 | BC087965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122086 | BC147461.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122086 | BC147464.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485436 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39902 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122086 | AK136484.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122086 | NM_001039720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122086 | BC087965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122086 | BC147461.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122086 | BC147464.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30342031 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08419461 | - | - |
| Whole Mouse Genome G4122A | A_52_P55258 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08419461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30341984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30341982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30341978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30341995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30341998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30341986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30341989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30341979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30341991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30342006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11910784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12077070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30341992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30341996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11910782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11958530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30341995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30341989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12282858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30341991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11958533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30341994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30341987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30341998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30342018 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10429589 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429591 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429590 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429592 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443889_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10429588 | - | - |
| Mouse Genome 430 2.0 Array | 1443889_at | - | - |
| Mouse Genome 430 2.0 Array | 1443889_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105892_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105892_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500013085.mm.1 | NR_108041.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013085.mm.1 | TC1500001736.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500013087.mm.1 | NR_108041.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013087.mm.1 | TC1500001736.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500013082.mm.1 | NR_108041.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013082.mm.1 | TC1500001736.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500013084.mm.1 | NR_108041.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013084.mm.1 | TC1500001736.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500013083.mm.1 | NR_108041.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500013083.mm.1 | TC1500001736.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1443889_at | - | - |
| Mouse Expression Set 430 | 1443889_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443889_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443889_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 766009 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214448 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3017818 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3017818 | - | - |

9030619P08Rik
Genomic Map