Assay Details
| Gene Symbol: Siglecf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271019.1 | NP_001257948.1 | 6 - 7 | 1083 | 68 | |
| NM_145581.2 | NP_663556.1 | 6 - 7 | 1083 | 68 | ||
| GenBank mRNA | AF293371.1 | - | 6 - 7 | 1019 | 68 | |
| AK146203.1 | - | 5 - 6 | 1120 | 68 | ||
| AY210401.1 | - | 5 - 6 | 979 | 68 | ||
| BC145038.1 | - | 5 - 6 | 1014 | 9053586 | 68 | |
| BC145987.1 | - | 5 - 6 | 1014 | 40131290 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Siglecf
Genomic Map