Assay Details
| Gene Symbol: Siglece | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031181.2 | NP_112458.2 | 1 - 2 | 467 | 121 | |
| GenBank mRNA | AF329269.1 | - | 1 - 2 | 471 | 121 | |
| AY348552.1 | - | 1 - 2 | 476 | 121 | ||
| AY371487.1 | - | 1 - 2 | 456 | 121 | ||
| BC023280.1 | - | 1 - 2 | 466 | 3662856 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Siglece
Genomic Map