Assay Details
| Gene Symbol: Siglece | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031181.2 | NP_112458.2 | 2 - 3 | 747 | 92 | |
| GenBank mRNA | AF329269.1 | - | 2 - 3 | 751 | 92 | |
More Information
Set Membership: |
|

Siglece
Genomic Map