Assay Details
| Gene Symbol: Siglece | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031181.2 | NP_112458.2 | 5 - 6 | 1153 | 62 | |
| GenBank mRNA | AF329269.1 | - | 5 - 6 | 1157 | 62 | |
| AY348552.1 | - | 5 - 6 | 1162 | 62 | ||
| AY371487.1 | - | 5 - 6 | 1142 | 62 | ||
| BC023280.1 | - | 5 - 6 | 1152 | 3662856 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Siglece
Genomic Map