Assay Details
| Gene Symbol: Tfip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018783.4 | NP_061253.2 | 6 - 7 | 945 | 92 | |
| GenBank mRNA | AF290474.3 | - | 6 - 7 | 945 | 92 | |
| AK147973.1 | - | 6 - 7 | 896 | 92 | ||
| AK148182.1 | - | 6 - 7 | 951 | 92 | ||
| AK148260.1 | - | 6 - 7 | 876 | 92 | ||
| AK161154.1 | - | 6 - 7 | 949 | 92 | ||
| BC017682.1 | - | 6 - 7 | 880 | 4239713 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tfip11
Genomic Map