Assay Details
| Gene Symbol: Zdhhc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178395.3 | NP_848482.1 | 10 - 11 | 1170 | 66 | |
| GenBank mRNA | AK030662.1 | - | 10 - 11 | 1171 | 66 | |
| AK046533.1 | - | 10 - 11 | 1170 | 66 | ||
| BC117761.1 | - | 11 - 12 | 932 | 66 | ||
| BC127156.1 | - | 11 - 12 | 1004 | 40090859 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc2
Genomic Map