Assay Details
| Gene Symbol: Zdhhc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178395.3 | NP_848482.1 | 7 - 8 | 823 | 114 | |
| GenBank mRNA | AK018284.1 | - | 114 | |||
| AK030662.1 | - | 7 - 8 | 824 | 114 | ||
| AK044652.1 | - | 7 - 8 | 817 | 114 | ||
| AK046533.1 | - | 7 - 8 | 823 | 114 | ||
| AK136296.1 | - | 7 - 8 | 822 | 114 | ||
| BC117761.1 | - | 8 - 9 | 585 | 114 | ||
| BC127156.1 | - | 8 - 9 | 657 | 40090859 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc2
Genomic Map