Assay Details
| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163144.1 | NP_001156616.1 | 10 - 11 | 1798 | 71 | |
| NM_001190483.1 | NP_001177412.1 | 10 - 11 | 1798 | 71 | ||
| GenBank mRNA | BC013068.1 | - | 10 - 11 | 1418 | 71 | |
| D12619.1 | - | 10 - 11 | 1387 | 71 | ||
| D17583.1 | - | 3 - 4 | 331 | 71 | ||
| L14932.1 | - | 10 - 11 | 1348 | 71 | ||
More Information
Set Membership: |
|

Pcsk5
Genomic Map