Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900010971.mm.1 |
NM_198019.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010971.mm.1 |
NM_198019.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010971.mm.1 |
TC1900001205.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010970.mm.1 |
NM_198019.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010970.mm.1 |
NM_198019.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010970.mm.1 |
TC1900001205.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010973.mm.1 |
NM_198019.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010973.mm.1 |
NM_198019.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010973.mm.1 |
TC1900001205.mm.1 |
MTA TC |