Assay Details
Gene Symbol: Cep78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198019.2 | NP_932136.2 | 9 - 10 | 1326 | 102 | |
GenBank mRNA | AK050870.1 | - | 8 - 9 | 1452 | 102 | |
AK160295.1 | - | 2 - 3 | 557 | 102 | ||
BC070400.1 | - | 9 - 10 | 1326 | 5694341 | 102 | |
BC071270.1 | - | 9 - 10 | 1308 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
