Assay Details
Gene Symbol: Olfr1423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146680.1 | NP_666891.1 | 1 | 883 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2060682 | NM_146680.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167043 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2060682 | NM_146680.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P159986 | NM_146680.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959218 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959203 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959193 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959208 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959213 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959216 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959219 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959207 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959217 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959201 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959210 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959198 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959172 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959188 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959194 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959191 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959171 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959206 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31959215 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167043 | - | - |
Whole Mouse Genome G4122A | A_51_P159986 | NM_146680.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114120 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167043 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14168892 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14308574 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14308585 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14308590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31959216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959209 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959163 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14328343 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959229 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959225 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959129 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959169 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959122 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14293566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14275424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14274125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959207 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14327722 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14228499 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14308583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14228507 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959131 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14150048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14446023 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959200 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14330386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14494500 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959128 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31959147 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10466277 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10466276 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 741564 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4560575 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4560575 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004560575 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246989 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1246989 | - | - |
mouse-6 v1.1 (Illumina) | scl52715.1.136_77-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4560575 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl52715.1.136_77-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560575 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1246989 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246989 | - | - |
