Assay Details
| Gene Symbol: Calhm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081271.1 | NP_001074740.1 | 1 - 2 | 552 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
cation transport
calcium ion transport
ATP transport
ion transmembrane transport
regulation of ion transmembrane transport
sensory perception of bitter taste
sensory perception of sweet taste
sensory perception of umami taste
protein homooligomerization
calcium ion transmembrane transport
cation transmembrane transport
calcium activated cation channel activity
voltage-gated ion channel activity
cation channel activity
calcium channel activity
identical protein binding
transport
ion transport
cation transport
calcium ion transport
ATP transport
ion transmembrane transport
regulation of ion transmembrane transport
sensory perception of bitter taste
sensory perception of sweet taste
sensory perception of umami taste
protein homooligomerization
calcium ion transmembrane transport
cation transmembrane transport
calcium activated cation channel activity
voltage-gated ion channel activity
cation channel activity
calcium channel activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178603 | NM_001081271.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863882 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178603 | NM_001081271.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32148028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32147987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14304974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14396955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14468835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14494413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14463223 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14255349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14396951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14396946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14376662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14466079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14304965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148031 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32148020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32147981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14485675 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10468301 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10468302 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10468300 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900014383.mm.1 | NM_001081271.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014383.mm.1 | NM_001081271.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014383.mm.1 | TC1900001624.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900014384.mm.1 | NM_001081271.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014384.mm.1 | NM_001081271.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900014384.mm.1 | TC1900001624.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 511476 | - | - |

Calhm1
Genomic Map