Assay Details
| Gene Symbol: Pdlim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016798.3 | NP_058078.1 | 4 - 5 | 617 | 68 | |
| GenBank mRNA | AF002283.1 | - | 4 - 5 | 641 | 68 | |
| AK016447.1 | - | 4 - 5 | 594 | 68 | ||
| AK078421.1 | - | 4 - 5 | 617 | 68 | ||
| BC070418.1 | - | 4 - 5 | 583 | 5714572 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdlim3
Genomic Map