Assay Details
Gene Symbol: Tlr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126166.4 | NP_569054.2 | 4 - 5 | 797 | 133 | |
GenBank mRNA | AF355152.1 | - | 2 - 3 | 479 | 133 | |
AF420279.1 | - | 1 - 2 | 436 | 133 | ||
AK083977.1 | - | 4 - 5 | 798 | 133 | ||
AK087739.1 | - | 3 - 4 | 619 | 133 | ||
AK166177.1 | - | 3 - 4 | 672 | 133 | ||
AK171438.1 | - | 4 - 5 | 779 | 133 | ||
BC099937.1 | - | 4 - 5 | 835 | 30084049 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
